CAS RN 75483-04-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Nrf2 Activator II, AI-1
The Nrf2 Activator II, AI-1, also referenced under CAS 75483-04-4, modulates the biological activity of Nrf2. This small molecule/inhibitor is primarily used for Biochemicals applications.