accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CH3CH3CH3C(E)

CAS RN 7549-37-3

IUPAC Name: (6E)-8,8-dimethoxy-2,6-dimethylocta-2,6-diene
Synonyms: citral dimethyl acetal
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: ZSKAJFSSXURRGL-PKNBQFBNSA-N
SMILES: COC(OC)\C=C(/C)CCC=C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results