accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOCH3(E/Z)(E/Z)

CAS RN 7549-44-2

IUPAC Name: methyl 3-[4-(3-methoxy-3-oxoprop-1-en-1-yl)phenyl]prop-2-enoate
Synonyms: methyl 3-[4-(3-methoxy-3-oxoprop-1-en-1-yl)phenyl]prop-2-enoate
Molecular Weight (g/mol): 246.26
Molecular Formula: C14H14O4
InChi Key: IFNSXAJHSAPYLB-UHFFFAOYSA-N
SMILES: COC(=O)C=CC1=CC=C(C=CC(=O)OC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results