CAS RN 7550-66-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Zinc Metal, Granular, 20-Mesh, Reagent Grade, Innovating Science™
CAS: 7550-66-6 Molecular Weight (g/mol): 65.38 g/mol
2
Zinc Metal Powder, 100-Mesh, Reagent Grade, Innovating Science™
CAS: 7550-66-6 Molecular Weight (g/mol): 65.38 g/mol
3
Zinc Metal Pieces, Lab Grade, Innovating Science™
CAS: 7550-66-6 Molecular Weight (g/mol): 65.38 g/mol
4
Zinc Metal Mossy, Reagent Grade, Innovating Science™
CAS: 7550-66-6 Molecular Weight (g/mol): 65.38 g/mol