accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHONOHNOHNClFFFF

CAS RN 755037-03-7

IUPAC Name: 4-[4-({[4-chloro-3-(trifluoromethyl)phenyl]carbamoyl}amino)-3-fluorophenoxy]-N-methylpyridine-2-carboxamide
Synonyms: regorafenib
Molecular Weight (g/mol): 482.82
Molecular Formula: C21H15ClF4N4O3
InChi Key: FNHKPVJBJVTLMP-UHFFFAOYSA-N
SMILES: CNC(=O)C1=NC=CC(OC2=CC(F)=C(NC(=O)NC3=CC(=C(Cl)C=C3)C(F)(F)F)C=C2)=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results