accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiLiLiH3COSHNOHNOH3CCH3OPOOHOPOHOOONNNNNH2OHOPOHOOHOH

CAS RN 75520-41-1

IUPAC Name: ({2-[({[({3-[(2-{[2-(acetylsulfanyl)ethyl]carbamoyl}ethyl)carbamoyl]-3-hydroxy-2,2-dimethylpropoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)methyl]-5-(6-amino-9H-purin-9-yl)-4-hydroxyoxolan-3-yl}oxy)phosphonic acid trilithium
Synonyms: Acetyl-CoA trilithium
Molecular Weight (g/mol): 830.39
Molecular Formula: C23H38Li3N7O17P3S
InChi Key: MOGXDJCKYPGZRO-UHFFFAOYNA-N
SMILES: [Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(O)C(C)(C)COP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1OP(O)(O)=O)N1C=NC2=C(N)N=CN=C12
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)