accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNO

CAS RN 7556-55-0

IUPAC Name: 4-methylpiperazine-1-carbaldehyde
Synonyms: 4-methylpiperazine-1-carbaldehyde
Molecular Weight (g/mol): 128.18
Molecular Formula: C6H12N2O
InChi Key: JQTMGOLZSBTZMS-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results