accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75568-11-5

IUPAC Name: 3,5-dibromobenzene-1,2-diamine
Synonyms: 3,5-dibromobenzene-1,2-diamine
Molecular Weight (g/mol): 265.94
Molecular Formula: C6H6Br2N2
InChi Key: BABDSAMUOCWCNS-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC(Br)=C1N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)