accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3(E/Z)

CAS RN 7560-43-2

IUPAC Name: methyl 3-(4-methylphenyl)prop-2-enoate
Synonyms: methyl 3-(4-methylphenyl)prop-2-enoate
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: WLJBRXRCJNSDHT-UHFFFAOYSA-N
SMILES: COC(=O)C=CC1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results