accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OH

CAS RN 7568-93-6

IUPAC Name: 2-amino-1-phenylethan-1-ol
Synonyms: β phenylethanolamine bisnorephedrine β-phenethanolamine 2-hydroxyphenethylamine 2-phenylethanolamine 2 hydroxyphenethylamine 2 phenylethanolamine phenylethanolamine phenethanolamine β-phenylethanolamine
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: ULSIYEODSMZIPX-UHFFFAOYNA-N
SMILES: NCC(O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)