accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7569-26-8

IUPAC Name: 1-chloromethanesulfonyl-4-methylbenzene
Synonyms: 1-chloromethanesulfonyl-4-methylbenzene
Molecular Weight (g/mol): 204.67
Molecular Formula: C8H9ClO2S
InChi Key: ZQPJNJKCPJDTIQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)CCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results