accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOH2COH3C

CAS RN 757-54-0

IUPAC Name: dimethyl (prop-2-en-1-yl)phosphonate
Synonyms: dimethyl prop-2-en-1-ylphosphonate
Molecular Weight (g/mol): 150.11
Molecular Formula: C5H11O3P
InChi Key: ZOSQAGGCVFVCNO-UHFFFAOYSA-N
SMILES: COP(=O)(CC=C)OC
Boiling Point 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results