accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NO

CAS RN 7570-45-8

IUPAC Name: 9-ethyl-9H-carbazole-3-carbaldehyde
Synonyms: 9-ethylcarbazole-3-carbaldehyde
Molecular Weight (g/mol): 223.28
Molecular Formula: C15H13NO
InChi Key: QGJXVBICNCIWEL-UHFFFAOYSA-N
SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)