accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNNH2

CAS RN 7570-49-2

IUPAC Name: 2-methyl-1H-indol-5-amine
Synonyms: 2-methyl-1H-indol-5-amine
Molecular Weight (g/mol): 146.19
Molecular Formula: C9H10N2
InChi Key: JQULCCZIXYRBSE-UHFFFAOYSA-N
SMILES: CC1=CC2=CC(N)=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results