accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7575-23-7

HSOOOOSHOOSHOOSH

CAS RN 7575-23-7

IUPAC Name: 3-[(3-sulfanylpropanoyl)oxy]-2,2-bis({[(3-sulfanylpropanoyl)oxy]methyl})propyl 3-sulfanylpropanoate
Synonyms: pentaerythrityl tetramercaptopropionate
Molecular Weight (g/mol): 488.64
Molecular Formula: C17H28O8S4
InChi Key: JOBBTVPTPXRUBP-UHFFFAOYSA-N
SMILES: SCCC(=O)OCC(COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results