accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOH

CAS RN 75754-04-0

IUPAC Name: 5-phenylpyridine-2-carboxylic acid
Synonyms: 5-phenylpyridine-2-carboxylic acid
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: CNCJXSKAKGXNKJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results