accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrHOOOHN

CAS RN 75776-67-9

IUPAC Name: 4-oxopyrrolidine-2-carboxylic acid hydrobromidyl
Synonyms: 4-oxoproline hydrobromidyl
Molecular Weight (g/mol): 209.02
Molecular Formula: C5H7BrNO3
InChi Key: BCIIQHDWHSKTHA-UHFFFAOYNA-N
SMILES: [Br].OC(=O)C1CC(=O)CN1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)