accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOO

CAS RN 7579-36-4

IUPAC Name: dibenzyl oxalate
Synonyms: dibenzyl oxalate
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: ZYZXGWGQYNTGAU-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results