accessibility menu, dialog, popup

UserName

Viewing profile as user:
NIF

CAS RN 757950-13-3

IUPAC Name: 3-fluoro-5-iodopyridine
Synonyms: 3-fluoro-5-iodopyridine
Molecular Weight (g/mol): 222.99
Molecular Formula: C5H3FIN
InChi Key: MMJSSIRVEYQWMU-UHFFFAOYSA-N
SMILES: FC1=CC(I)=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results