accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNBrI

CAS RN 757978-18-0

IUPAC Name: 5-bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine
Synonyms: 5-bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 322.93
Molecular Formula: C7H4BrIN2
InChi Key: GIPGJYARDOQGDJ-UHFFFAOYSA-N
SMILES: BrC1=CN=C2NC=C(I)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results