accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 758-12-3

IUPAC Name: acetate
Synonyms: acetate caswell no. 003 glacial acetate vinegar (substance) azetat acetic acid product (product) fema number 2006 ethanoat acetic acid,glacial ion(1-) acetic acid
Molecular Weight (g/mol): 59.05
Molecular Formula: C2H3O2
InChi Key: QTBSBXVTEAMEQO-UHFFFAOYSA-M
SMILES: CC([O-])=O
Grade 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)