accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7580-67-8

IUPAC Name: lithium(1+) hydride
Synonyms: lithium hydride
Molecular Weight (g/mol): 7.95
Molecular Formula: HLi
InChi Key: SRTHRWZAMDZJOS-UHFFFAOYSA-N
SMILES: [H-].[Li+]
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)