accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75806-84-7

IUPAC Name: 2-bromo-3-chloro-5-(trifluoromethyl)pyridine
Synonyms: 2-bromo-3-chloro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 260.44
Molecular Formula: C6H2BrClF3N
InChi Key: SMTKGMYGLYWNDL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=C(Br)N=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)