accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClNBr

CAS RN 75806-86-9

IUPAC Name: 2-bromo-5-chloro-3-nitropyridine
Synonyms: 2-bromo-5-chloro-3-nitropyridine
Molecular Weight (g/mol): 237.44
Molecular Formula: C5H2BrClN2O2
InChi Key: VFMIDVIAQMFGGP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=CN=C1Br
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)