accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7583-27-9

CH3ICH3I

CAS RN 7583-27-9

IUPAC Name: 4,4'-diiodo-3,3'-dimethyl-1,1'-biphenyl
Synonyms: 4,4'-diiodo-3,3'-dimethyl-1,1'-biphenyl
Molecular Weight (g/mol): 434.06
Molecular Formula: C14H12I2
InChi Key: ISWXEMYZGWXIIZ-UHFFFAOYSA-N
SMILES: CC1=C(I)C=CC(=C1)C1=CC(C)=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results