accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75867-46-8

IUPAC Name: 2,6-diethynylpyridine
Synonyms: 2,6-diethynylpyridine
Molecular Weight (g/mol): 127.15
Molecular Formula: C9H5N
InChi Key: VYRLFYTZNXGQIK-UHFFFAOYSA-N
SMILES: C#CC1=CC=CC(=N1)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)