accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHF

CAS RN 7589-27-7

IUPAC Name: 2-(4-fluorophenyl)ethan-1-ol
Synonyms: 4-fluorophenethyl alcohol
Molecular Weight (g/mol): 140.16
Molecular Formula: C8H9FO
InChi Key: MWUVGXCUHWKQJE-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)