CAS RN 75894-99-4
IUPAC Name:
1,3-dibromo-5-octylbenzene
Synonyms:
1,3-dibromo-5-octylbenzene
Molecular Weight (g/mol):
348.12
Molecular Formula:
C14H20Br2
InChi Key:
OHOXSSBAVJERNK-UHFFFAOYSA-N
SMILES:
CCCCCCCCC1=CC(Br)=CC(Br)=C1
1
–
1
of
1
results
1
1,3-Dibromo-5-n-octylbenzene 98.0+%, TCI America™
CAS: 75894-99-4 Molecular Formula: C14H20Br2 Molecular Weight (g/mol): 348.122 InChI Key: OHOXSSBAVJERNK-UHFFFAOYSA-N PubChem CID: 101748485 IUPAC Name: 1,3-dibromo-5-octylbenzene SMILES: CCCCCCCCC1=CC(=CC(=C1)Br)Br