accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75898-34-9

IUPAC Name: 2'-nitro-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 2'-nitro-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 224.22
Molecular Formula: C13H8N2O2
InChi Key: LZUDBRIMDISCJM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results