accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 759-65-9

IUPAC Name: 1,4-diethyl 2-methyl-3-oxobutanedioate
Synonyms: diethyl oxalpropionate ethyl α-ethoxalylpropionate
Molecular Weight (g/mol): 202.21
Molecular Formula: C9H14O5
InChi Key: OQOCQBJWOCRPQY-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)C(=O)C(=O)OCC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results