CAS RN 759-94-4
IUPAC Name:
N,N-dipropyl(ethylsulfanyl)formamide
Synonyms:
torbin caswell no. 435 genep eptc C9H19nos EPTC stauffer R 1608 niptan epa pesticide chemical code 041401 alirox eptc (herbicide)
Molecular Weight (g/mol):
189.32
Molecular Formula:
C9H19NOS
InChi Key:
GUVLYNGULCJVDO-UHFFFAOYSA-N
SMILES:
CCCN(CCC)C(=O)SCC
1
–
1
of
1
results
1
EPTC, SPEX CertiPrep™
CAS: 759-94-4 Molecular Formula: C9H19NOS Molecular Weight (g/mol): 189.317 InChI Key: GUVLYNGULCJVDO-UHFFFAOYSA-N PubChem CID: 12968 ChEBI: CHEBI:4738 IUPAC Name: S-ethyl N,N-dipropylcarbamothioate SMILES: CCCN(CCC)C(=O)SCC