CAS RN 75921-69-6
IUPAC Name:
(4S)-4-{[(1S)-1-{[(1R)-1-{[(1S)-1-{[(1S)-1-[({[(2S)-6-amino-1-[(2S)-2-{[(1S)-1-carbamoyl-2-methylpropyl]carbamoyl}pyrrolidin-1-yl]-1-oxohexan-2-yl]carbamoyl}methyl)carbamoyl]-2-(1H-indol-3-yl)ethyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-2-phenylethyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl]carbamoyl}-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-acetamido-3-hydroxypropanamido]-3-(4-hydroxyphenyl)propanamido]-3-hydroxypropanamido]hexanamido]butanoic acid
Synonyms:
afamelanotide
Molecular Weight (g/mol):
1646.87
Molecular Formula:
C78H111N21O19
InChi Key:
UAHFGYDRQSXQEB-LEBBXHLNSA-N
SMILES:
CCCC[C@H](NC(=O)[C@H](CO)NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@H](CO)NC(C)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N[C@H](CC1=CC=CC=C1)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C(C)C)C(N)=O
1
–
1
of
1
results
1
[Nle4, D-Phe7] alpha-Melanocyte Stimulating Hormone amide trifluoroacetate salt, Thermo Scientific Chemicals
CAS: 75921-69-6 Molecular Formula: C78H111N21O19 MDL Number: MFCD00151715 Synonym: N-Acetyl-Ser-Tyr-Ser-Nle-Glu-His-DPhe-Arg-Trp-Gly-Lys-Pro-Val-NH{2},Melanotan