accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75932-02-4

IUPAC Name: (2S)-2,6-bis({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-2,6-bis({[(9H-fluoren-9-ylmethoxy)carbonyl]amino})hexanoic acid|FMOC-D-LYS(FMOC)-OH
Molecular Weight (g/mol): 590.68
Molecular Formula: C36H34N2O6
InChi Key: BMJRTKDVFXYEFS-YGFIFORBNA-N
SMILES: OC(=O)[C@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)