accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7596-80-7

IUPAC Name: N-(2-methanesulfonamidophenyl)methanesulfonamide
Synonyms: N-(2-methanesulfonamidophenyl)methanesulfonamide
Molecular Weight (g/mol): 264.31
Molecular Formula: C8H12N2O4S2
InChi Key: BYZJRLRCNOECEV-UHFFFAOYSA-N
SMILES: CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results