accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7598-61-0

IUPAC Name: diethyl (2,2-diethoxyethyl)phosphonate
Synonyms: diethyl 2,2-diethoxyethylphosphonate
Molecular Weight (g/mol): 254.26
Molecular Formula: C10H23O5P
InChi Key: LUQYELQXRPNKRY-UHFFFAOYSA-N
SMILES: CCOC(CP(=O)(OCC)OCC)OCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)