accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75980-60-8

IUPAC Name: (diphenylphosphoroso)(2,4,6-trimethylphenyl)methanone
Synonyms: trimethylbenzoyl diphenylphosphine oxide
Molecular Weight (g/mol): 348.38
Molecular Formula: C22H21O2P
InChi Key: VFHVQBAGLAREND-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(=O)P(=O)(C2=CC=CC=C2)C2=CC=CC=C2)C(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results