accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 75996-29-1

IUPAC Name: 5-fluorobenzene-1,3-diol
Synonyms: 5-fluorobenzene-1,3-diol
Molecular Weight (g/mol): 128.1
Molecular Formula: C6H5FO2
InChi Key: LYFBZGKZAZBANN-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results