CAS RN 76-03-9
Trichloroacetic acid, ACS, 99% min
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 MDL Number: MFCD00004177 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic acid, 99%, extra pure
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.39 MDL Number: MFCD00004177 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic acid, 99+%, ACS reagent
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 MDL Number: MFCD00004177 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic Acid, Fisher Chemical
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 MDL Number: MFCD00004177 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic Acid (Crystalline/Certified ACS), Fisher Chemical™
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 MDL Number: MFCD00004177 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic Acid (Crystalline/Certified), Fisher Chemical™
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 MDL Number: MFCD00004177 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic Acid, 80% (w/v), Ricca Chemical
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): Mixture InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: trichloroacetic acid SMILES: OC(=O)C(Cl)(Cl)Cl
Trichloroacetic acid. EMSURE™, MilliporeSigma™, For Analysis,
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic acid, ACS, ≥99%, MP Biomedicals™
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic acid, 99.6%, MP Biomedicals™
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Solstice Detritylation Reagent, 3%Trichloroacetic Acid in Dichloromethane (withv)
BioSyn™, for DNA/RNA synthesis
Trichloroacetic Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic acid, 99%
CAS: 76-03-9 Molecular Formula: C2HCl3O2 Molecular Weight (g/mol): 163.378 MDL Number: MFCD00004177 InChI Key: YNJBWRMUSHSURL-UHFFFAOYSA-N Synonym: trichloroacetic acid,trichloroethanoic acid,aceto-caustin,trichloroacetic acid,acetic acid, trichloro,trichloressigsaeure,konesta,acide trichloracetique,acido tricloroacetico,amchem grass killer PubChem CID: 6421 ChEBI: CHEBI:30956 IUPAC Name: 2,2,2-trichloroacetic acid SMILES: C(=O)(C(Cl)(Cl)Cl)O
Trichloroacetic Acid, 20% (w/v) Aqueous Solution, Spectrum™ Chemical
CAS: 76-03-9
Trichloroacetic Acid, 50% (w/v) Aqueous Solution, Spectrum™ Chemical
CAS: 76-03-9