accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76-12-0

IUPAC Name: 1,1,2,2-tetrachloro-1,2-difluoroethane
Synonyms: 1,1,2,2-tetrachloro-1,2-difluoroethane
Molecular Weight (g/mol): 203.82
Molecular Formula: C2Cl4F2
InChi Key: UGCSPKPEHQEOSR-UHFFFAOYSA-N
SMILES: FC(Cl)(Cl)C(F)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results