accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOFFFF

CAS RN 76-37-9

IUPAC Name: 2,2,3,3-tetrafluoropropan-1-ol
Synonyms: 2,2,3,3-tetrafluoropropanol
Molecular Weight (g/mol): 132.06
Molecular Formula: C3H4F4O
InChi Key: NBUKAOOFKZFCGD-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)F
Boiling Point 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)