accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3HOOSOOCH3H3COHCH3

CAS RN 76-61-9

IUPAC Name: 3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: thymol blue
Molecular Weight (g/mol): 466.59
Molecular Formula: C27H30O5S
InChi Key: PRZSXZWFJHEZBJ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=C(C)C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
Molecular Weight (g/mol) 
  • (16)
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (5)
  • (2)
  • (1)
Grade 
  • (8)
  • (3)
  • (1)
  • (2)
  • (3)
Color 
  • (6)
  • (6)
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (5)
  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (11)
  • (16)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (5)
  • (2)
  • (1)
  • (8)
  • (3)
  • (1)
  • (2)
  • (3)
  • (6)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)