accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76-63-1

IUPAC Name: triphenyl(prop-2-en-1-yl)stannane
Synonyms: stannane, allyltriphenyl-
Molecular Weight (g/mol): 391.1
Molecular Formula: C21H20Sn
InChi Key: NDUYAGLANMHJHF-UHFFFAOYSA-N
SMILES: C=CC[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results