accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76-66-4

IUPAC Name: methyl (2E)-2-[(3R,6'R,7'S,8'aS)-6'-ethyl-2-oxo-1,2,3',5',6',7',8',8'a-octahydro-2'H-spiro[indole-3,1'-indolizin]-7'-yl]-3-methoxyprop-2-enoate
Synonyms: rhynchophylline
Molecular Weight (g/mol): 384.48
Molecular Formula: C22H28N2O4
InChi Key: DAXYUDFNWXHGBE-KAXDATADSA-N
SMILES: CC[C@H]1CN2CC[C@@]3([C@@H]2C[C@@H]1\C(=C/OC)C(=O)OC)C(=O)NC1=CC=CC=C31
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results