CAS RN 76-73-3
CAS RN 76-73-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma Supelco CERILLIANT™ Secobarbital Solution, 1 mg/mL in methanol, Sold by MilliporeSigma™ Supelco™
This Certified Spiking Solution™ is suitable for numerous GC/MS or LC/MS testing applications from clinical toxicology to urine drug testing and forensic analysis.