accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSi

CAS RN 76-86-8

IUPAC Name: chlorotriphenylsilane
Synonyms: chlorotriphenylsilane
Molecular Weight (g/mol): 294.85
Molecular Formula: C18H15ClSi
InChi Key: MNKYQPOFRKPUAE-UHFFFAOYSA-N
SMILES: Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (2)
  • (4)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (4)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (8)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)