accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 760-21-4

IUPAC Name: 3-methylidenepentane
Synonyms: 2-ethyl-1-butene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: RYKZRKKEYSRDNF-UHFFFAOYSA-N
SMILES: CCC(=C)CC
Boiling Point 
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)