accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 760-67-8

IUPAC Name: 2-ethylhexanoyl chloride
Synonyms: hexanoyl chloride, 2-ethyl-
Molecular Weight (g/mol): 162.66
Molecular Formula: C8H15ClO
InChi Key: WFSGQBNCVASPMW-UHFFFAOYNA-N
SMILES: CCCCC(CC)C(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results