accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2OHO

CAS RN 760-78-1

IUPAC Name: 2-aminopentanoic acid
Synonyms: Nva|DL-norvaline|norvalines|2-aminopentanoic acids|2-aminovaleric acids|α-DL-aminopentanoic acid
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: SNDPXSYFESPGGJ-UHFFFAOYSA-N
SMILES: CCCC(N)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)