accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7601-54-9

IUPAC Name: trisodium phosphate
Synonyms: sodium phosphate tertiary sodium phosphate tertiaeres natriumphosphat(V) tribasic sodium orthophosphate Na3PO4 phosphoric acid trisodium salt trinatriumphosphat tribasic sodium phosphate sodium phosphate, tribasic sodium orthophosphate
Molecular Weight (g/mol): 163.94
Molecular Formula: Na3O4P
InChi Key: RYFMWSXOAZQYPI-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (14)
Quantity 
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
Grade 
  • (4)
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (4)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (8)
  • (5)
  • (4)
  • (2)
  • (14)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (3)
  • (6)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)