CAS RN 7601-90-3
Perchloric Acid Solution, N/10 (0.1N) (Certified), Fisher Chemical™
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric Acid, 70% (Certified ACS), Fisher Chemical™
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric acid, 0.1N in Acetic Acid Standardized Solution
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric acid, ACS reagent, ca. 70% solution in water
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric acid, pure, 0.1M solution in acetic acid
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Perchloric acid, for analysis, ca. 70% solution in water
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric Acid, 60% (Certified ACS), Fisher Chemical™
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric Acid (Optima™), Fisher Chemical™
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric Acid (TraceMetal™ Grade), Fisher Chemical™
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric acid, ACS, 48-50%
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric acid, ACS, 69.0-72.0%, redistilled
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric Acid, Normality about 12, 69 to 72%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric Acid, 60 to 62%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O
Perchloric Acid, 69 to 72%, BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™
CAS: 7601-90-3 Molecular Formula: ClHO4 Molecular Weight (g/mol): 100.45 MDL Number: MFCD00011325 InChI Key: VLTRZXGMWDSKGL-UHFFFAOYSA-N Synonym: unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchloric acid,ueberchlorsaeure,hyperchloric acid,hclo4 PubChem CID: 24247 ChEBI: CHEBI:29221 IUPAC Name: perchloric acid SMILES: O[Cl](=O)(=O)=O